Merck
  • Profile of polychlorinated biphenyls in the brown alga Padina sanctae-crucis along the Orote dump coastline, Orote Peninsula, Western Guam.
Profile of polychlorinated biphenyls in the brown alga Padina sanctae-crucis along the Orote dump coastline, Orote Peninsula, Western Guam.
Journal of toxicology and environmental health. Part A (2010-09-08)
Brian Christopher Schaible
ABSTRACT
The purpose of this study was to determine whether the brown seaweed Padina sanctae-crucis might be used as a biomonitor organism to determine the location, distribution, and type of polychlorinated biphenyls (PCB) present along a contaminated tropical coastline. Polychlorinated biphenyls were detected in P. sanctae-crucis collected from near-shore waters along a 4-km stretch of coastline. The profile of PCB levels from samples obtained at 13 sites along the coastline demonstrated a positive concentration gradient as sample locations progressed toward the dump site. Sample locations nearest the dump site revealed PCB levels twofold higher than background levels for P. sanctae-crucis. Chromatograms of samples obtained nearest the dump site indicated a similarity to the Aroclor 1260 fingerprint. The P. sanctae-crucis frond trimmings used for laboratory analysis were approximately 3-5 wk old. This study was conducted 8 mo following the completion of the removal actions at the dump site. Data suggest a release of Aroclor 1260 into the marine environment approximately 7 mo following the completion of removal actions at the dump site.
Product Description
Supelco
Aroclor 1260 solution, certified reference material, 200 μg/mL in methanol
Supelco
Aroclor 1260 solution, certified reference material, 500 mg/kg in transformer oil