Merck
  • Potent memapsin 2 (beta-secretase) inhibitors: design, synthesis, protein-ligand X-ray structure, and in vivo evaluation.
Potent memapsin 2 (beta-secretase) inhibitors: design, synthesis, protein-ligand X-ray structure, and in vivo evaluation.
Bioorganic & medicinal chemistry letters (2008-01-09)
Arun K Ghosh, Nagaswamy Kumaragurubaran, Lin Hong, Sarang Kulkarni, Xiaoming Xu, Heather B Miller, Dandepally Srinivasa Reddy, Vajira Weerasena, Robert Turner, Wanpin Chang, Gerald Koelsch, Jordan Tang
ABSTRACT
Structure-based design, synthesis, and biological evaluation of a series of peptidomimetic beta-secretase inhibitors incorporating hydroxyethylamine isosteres are described. We have identified inhibitor 24 which has shown exceedingly potent activity in memapsin 2 enzyme inhibitory (K(i) 1.8 nM) and cellular (IC(50)=1 nM in Chinese hamster ovary cells) assays. Inhibitor 24 has also shown very impressive in vivo properties (up to 65% reduction of plasma A beta) in transgenic mice. The X-ray structure of protein-ligand complex of memapsin 2 revealed critical interactions in the memapsin 2 active site.
Sigma-Aldrich
Leu-Ala hydrate, Bulk package