Merck
  • Preparation and characterization of a self-emulsifying pellet formulation.
Preparation and characterization of a self-emulsifying pellet formulation.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2007-01-02)
Ahmed Abdalla, Karsten Mäder
ABSTRACT
The purpose of the current study is to investigate the feasibility of producing solid self-emulsifying pellets using the extrusion/spheronization technique. Pellets were made from a mixture of C18 partial glycerides, Solutol HS15 and microcrystalline cellulose. Pellets with good physical properties (size, shape, friability) and self-emulsifying properties were produced. The pellets were, in contrast to pellets lacking Solutol, able to transfer a lipophilic dye and a spin probe into the aqueous media. The release kinetics and the microenvironment of the pellets during the release process were assessed using electron spin resonance (ESR) spectroscopy. The ESR results showed that the hydrophobic spin probe was localized mainly in the lipid environment all over the release time. Furthermore, the formulation was capable of accelerating the release of the drug diazepam and achieving a diazepam concentration above its saturation solubility. In conclusion, spherical pellets with low friability and self-emulsifying properties can be produced by the standard extrusion/spheronization technique. The pellets are capable of transfering lipophilic compounds into the aqueous phase and have a high potential to increase the bioavailability of lipophilic drugs.
Product Description
Supelco
Sudan Red 7B, analytical standard
Sigma-Aldrich
Sudan Red 7B
Sigma-Aldrich
Sudan Red 7B, for microscopy (Bot., Hist.)