Merck
Enzymatic degradation of chlorodiamino-s-triazine.
Applied and environmental microbiology (2002-08-30)
Jennifer L Seffernick, Nir Shapir, Michael Schoeb, Gilbert Johnson, Michael J Sadowsky, Lawrence P Wackett
ABSTRACT
2-Chloro-4,6-diamino-s-triazine (CAAT) is a metabolite of atrazine biodegradation in soils. Atrazine chlorohydrolase (AtzA) catalyzes the dechlorination of atrazine but is unreactive with CAAT. In this study, melamine deaminase (TriA), which is 98% identical to AtzA, catalyzed deamination of CAAT to produce 2-chloro-4-amino-6-hydroxy-s-triazine (CAOT). CAOT underwent dechlorination via hydroxyatrazine ethylaminohydrolase (AtzB) to yield ammelide. This represents a newly discovered dechlorination reaction for AtzB. Ammelide was subsequently hydrolyzed by N-isopropylammelide isopropylaminohydrolase to produce cyanuric acid, a compound metabolized by a variety of soil bacteria.
Product Description
Supelco
2-Chloro-4,6-diamino-1,3,5-triazine, analytical standard
Sigma-Aldrich
2-Chloro-4,6-diamino-1,3,5-triazine, 95%
Supelco
Ammelide, analytical standard