Merck
  • Metabolic clogging of mannose triggers dNTP loss and genomic instability in human cancer cells.
Metabolic clogging of mannose triggers dNTP loss and genomic instability in human cancer cells.
eLife (2023-07-18)
Yoichiro Harada, Yu Mizote, Takehiro Suzuki, Akiyoshi Hirayama, Satsuki Ikeda, Mikako Nishida, Toru Hiratsuka, Ayaka Ueda, Yusuke Imagawa, Kento Maeda, Yuki Ohkawa, Junko Murai, Hudson H Freeze, Eiji Miyoshi, Shigeki Higashiyama, Heiichiro Udono, Naoshi Dohmae, Hideaki Tahara, Naoyuki Taniguchi
ABSTRACT
Mannose has anticancer activity that inhibits cell proliferation and enhances the efficacy of chemotherapy. How mannose exerts its anticancer activity, however, remains poorly understood. Here, using genetically engineered human cancer cells that permit the precise control of mannose metabolic flux, we demonstrate that the large influx of mannose exceeding its metabolic capacity induced metabolic remodeling, leading to the generation of slow-cycling cells with limited deoxyribonucleoside triphosphates (dNTPs). This metabolic remodeling impaired dormant origin firing required to rescue stalled forks by cisplatin, thus exacerbating replication stress. Importantly, pharmacological inhibition of de novo dNTP biosynthesis was sufficient to retard cell cycle progression, sensitize cells to cisplatin, and inhibit dormant origin firing, suggesting dNTP loss-induced genomic instability as a central mechanism for the anticancer activity of mannose.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
VE-821, ≥98% (HPLC)