Merck
  • Anti-apoptotic effect of adipose tissue-derived stromal vascular fraction in denervated rat muscle.
Anti-apoptotic effect of adipose tissue-derived stromal vascular fraction in denervated rat muscle.
Stem cell research & therapy (2021-03-06)
R El-Habta, G Andersson, P J Kingham, L J Backman
ABSTRACT
Recovery of muscle function after peripheral nerve injury is often poor, and this can be attributed to muscle fiber atrophy and cell death. In the current study, we have investigated the effects of stromal vascular fraction (SVF) on muscle cell apoptosis and its potential to preserve muscle tissue following denervation. Rat gastrocnemius muscle was denervated by sciatic nerve transection. At 2 and 4 weeks after injury, muscles were examined histologically and apoptosis was measured using TUNEL assay and PCR array for a range of apoptotic genes. Additionally, an in vitro TNF-α apoptosis model was established using SVF cells co-cultured indirectly with primary rat myoblasts. Annexin V and TUNEL were used together with Western blotting to investigate the signaling pathways. Denervated muscles showed significantly higher TUNEL reactivity at 2 and 4 weeks following nerve injury, and an increased expression of caspase family genes, mitochondria-related apoptotic genes, and tumor necrosis factor family genes. In cultured rat primary myoblasts, Annexin V labeling was significantly increased at 12 h after TNF-α treatment, and this was followed by a significant increase in TUNEL reactivity at 48 h. Western blotting showed that caspase-7 was activated/cleaved as well as the downstream substrate, poly (ADP-ribose) polymerase (PARP). Co-culture of myoblasts with SVF significantly reduced all these measures of apoptosis. Bax and Bcl-2 levels were not changed suggesting that the TNF-α-induced apoptosis occurred via mitochondria-independent pathways. The protective effect of SVF was also shown in vivo; injections of SVF cells into denervated muscle significantly improved the mean fiber area and diameter, as well as reduced the levels of TUNEL reactivity. This study provides new insights into how adipose tissue-derived cells might provide therapeutic benefits by preserving muscle tissue.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Supelco
Methyl behenate, analytical standard
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution