Merck
  • Dynamic Covalent Chemistry under High-Pressure:A New Route to Disulfide Metathesis.
Dynamic Covalent Chemistry under High-Pressure:A New Route to Disulfide Metathesis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2018-04-21)
Szymon Sobczak, Wojciech Drożdż, Giulio I Lampronti, Ana M Belenguer, Andrzej Katrusiak, Artur R Stefankiewicz
ABSTRACT
This work describes, for the first time, the application of combined pressure and temperature stimuli in disulfide metathesis reactions. In the system studied, above a pressure of 0.2 GPa, equimolar amounts of symmetric disulfides bis 4-chlorophenyl disulfide [(4-ClPhS)2 ] and bis 2-nitrophenyl disulfide [(2-NO2 PhS)2 ] react to give the heterodimeric product 4-Cl-PhSSPh-2-NO2 . In contrast to experiments conducted in solution at atmospheric pressure or in mechanochemical experiments under ball-mill grinding conditions, there is no necessity to use a base or thiolate anion as a catalyst for the exchange reaction under investigated conditions. Single-crystal and powder X-ray diffraction revealed also that, despite the high-pressure conditions of this reaction, the heterodimeric-disulfide product unexpectedly crystallizes into the low-density polymorph A. This counterintuitive result contrasts with the high-pressure stability of the higher-density polymorph B, confirmed by its compression up to 2.8 GPa with no signs of a phase transition.
Sigma-Aldrich
Bis(2-nitrophenyl) disulfide, 99%
Sigma-Aldrich
Bis(4-chlorophenyl) disulfide, 97%