Merck
  • Rack1 Controls Parallel Fiber-Purkinje Cell Synaptogenesis and Synaptic Transmission.
Rack1 Controls Parallel Fiber-Purkinje Cell Synaptogenesis and Synaptic Transmission.
Frontiers in cellular neuroscience (2020-01-11)
Haihong Yang, Chaojuan Yang, Qian Zhu, Mengping Wei, Ying Li, Juanxian Cheng, Fengjiao Liu, Yan Wu, Jiyan Zhang, Chen Zhang, Haitao Wu
ABSTRACT
Purkinje cells (PCs) in the cerebellum receive two excitatory afferents including granule cells-derived parallel fiber (PF) and the climbing fiber. Scaffolding protein Rack1 is highly expressed in the cerebellar PCs. Here, we found delayed formation of specific cerebellar vermis lobule and impaired motor coordination in PC-specific Rack1 conditional knockout mice. Our studies further revealed that Rack1 is essential for PF-PC synapse formation. In addition, Rack1 plays a critical role in regulating synaptic plasticity and long-term depression (LTD) induction of PF-PC synapses without changing the expression of postsynaptic proteins. Together, we have discovered Rack1 as the crucial molecule that controls PF-PC synaptogenesis and synaptic plasticity. Our studies provide a novel molecular insight into the mechanisms underlying the neural development and neuroplasticity in the cerebellum.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-RACK1 (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid