Merck
  • A Sialylated Voltage-Dependent Ca2+ Channel Binds Hemagglutinin and Mediates Influenza A Virus Entry into Mammalian Cells.
A Sialylated Voltage-Dependent Ca2+ Channel Binds Hemagglutinin and Mediates Influenza A Virus Entry into Mammalian Cells.
Cell host & microbe (2018-05-22)
Yoichiro Fujioka, Shinya Nishide, Toyoyuki Ose, Tadaki Suzuki, Izumi Kato, Hideo Fukuhara, Mari Fujioka, Kosui Horiuchi, Aya O Satoh, Prabha Nepal, Sayaka Kashiwagi, Jing Wang, Mika Horiguchi, Yuko Sato, Sarad Paudel, Asuka Nanbo, Tadaaki Miyazaki, Hideki Hasegawa, Katsumi Maenaka, Yusuke Ohba
ABSTRACT
Influenza A virus (IAV) infection is initiated by the attachment of the viral glycoprotein hemagglutinin (HA) to sialic acid on the host cell surface. However, the sialic acid-containing receptor crucial for IAV infection has remained unidentified. Here, we show that HA binds to the voltage-dependent Ca2+ channel Cav1.2 to trigger intracellular Ca2+ oscillations and subsequent IAV entry and replication. IAV entry was inhibited by Ca2+ channel blockers (CCBs) or by knockdown of Cav1.2. The CCB diltiazem also inhibited virus replication in vivo. Reintroduction of wild-type but not the glycosylation-deficient mutants of Cav1.2 restored Ca2+ oscillations and virus infection in Cav1.2-depleted cells, demonstrating the significance of Cav1.2 sialylation. Taken together, we identify Cav1.2 as a sialylated host cell surface receptor that binds HA and is critical for IAV entry.
Sigma-Aldrich
Amlodipine besylate, ≥98% (HPLC)
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)