Merck
  • Loss of KLHL6 promotes diffuse large B-cell lymphoma growth and survival by stabilizing the mRNA decay factor roquin2.
Loss of KLHL6 promotes diffuse large B-cell lymphoma growth and survival by stabilizing the mRNA decay factor roquin2.
Nature cell biology (2018-04-27)
Jaewoo Choi, Kyutae Lee, Kristin Ingvarsdottir, Roberto Bonasio, Anita Saraf, Laurence Florens, Michael P Washburn, Saber Tadros, Michael R Green, Luca Busino
ABSTRACT
Kelch-like protein 6 (KLHL6) is an uncharacterized gene mutated in diffuse large B-cell lymphoma (DLBCL). Here we report that KLHL6 assembles with cullin3 to form a functional cullin-RING ubiquitin ligase. Mutations in KLHL6 inhibit its ligase activity by disrupting the interaction with cullin3. Loss of KLHL6 favours DLBCL growth and survival both in vitro and in xenograft models. We further established that the mRNA decay factor roquin2 is a substrate of KLHL6. Degradation of roquin2 is dependent on B-cell receptor activation, and requires the integrity of the Tyr691 residue in roquin2 that is essential for its interaction with KLHL6. A non-degradable roquin2(Y691F) mutant requires its RNA-binding ability to phenocopy the effect of KLHL6 loss. Stabilization of roquin2 promotes mRNA decay of the tumour suppressor and NF-κB pathway inhibitor, tumour necrosis factor-α-inducible gene 3. Collectively, our findings uncover the tumour suppressing mechanism of KLHL6.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H2A (acidic patch) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Roquin-1/2, clone 3F12 Antibody, clone 3F12, from rat
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse