![Xylitol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/237/059/59408280-d5c3-4b90-89bd-9cd78b45d6a2/640/59408280-d5c3-4b90-89bd-9cd78b45d6a2.png)
Xylitol
![Xylitol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/237/059/59408280-d5c3-4b90-89bd-9cd78b45d6a2/640/59408280-d5c3-4b90-89bd-9cd78b45d6a2.png)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing