
Tilmicosin

Synonym(s):
20-Deoxo-20-(3,5-dimethyl-1-piperidinyl)desmycosin
Empirical Formula (Hill Notation):
C46H80N2O13
Molecular Weight:
869.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3