
Tibolone for system suitability

Synonym(s):
Tibolone, (7α,17α)-17-Hydroxy-7-methyl-19-norpregn-5(10)-en-20-yn-3-one
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing