![Tetrakis(dimethylamido)hafnium(IV) packaged for use in deposition systems](/deepweb/assets/sigmaaldrich/product/structures/495/907/4d4d9d40-beab-4408-a249-28163802714b/640/4d4d9d40-beab-4408-a249-28163802714b.png)
Tetrakis(dimethylamido)hafnium(IV)
![Tetrakis(dimethylamido)hafnium(IV) packaged for use in deposition systems](/deepweb/assets/sigmaaldrich/product/structures/495/907/4d4d9d40-beab-4408-a249-28163802714b/640/4d4d9d40-beab-4408-a249-28163802714b.png)
Synonym(s):
TDMAH, Tetrakis(dimethylamino)hafnium(IV)
Linear Formula:
[(CH3)2N]4Hf
CAS Number:
Molecular Weight:
354.79
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing