
Temazepam impurity G

Synonym(s):
(3RS)-7-Chloro-1,4-dimethyl-5-phenyl-4,5-dihydro-1H-1,4-benzodiazepin-2,3-dione
Empirical Formula (Hill Notation):
C17H15ClN2O2
CAS Number:
Molecular Weight:
314.77
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing