
Tectoridin

Synonym(s):
D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, Shekanin, Tectorigenin 7-O-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C22H22O11
CAS Number:
Molecular Weight:
462.40
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing