
Sultamicillin for peak identification

Synonym(s):
Sultamicillin
Empirical Formula (Hill Notation):
C25H30N4O9S2
CAS Number:
Molecular Weight:
594.66
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing