
Silylating mixture I according to Sweeley

Synonym(s):
Chlorotrimethylsilane – hexamethyldisilazane mixture, HMDS + TMCS
CAS Number:
Products
Product Category
Brand
Physical Form
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing