
Rivastigmine Related Compound C

Synonym(s):
(S)-3-[1-(Dimethylamino)ethyl]phenol
Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2