
Puerarin

Synonym(s):
8-(β-D-Glucopyranosyl-7-hydroxy-3- (4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kakonein
Empirical Formula (Hill Notation):
C21H20O9
CAS Number:
Molecular Weight:
416.38
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing