
Ponceau S solution

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing