Skip to Content
Merck
Poly(ethylene glycol) methyl ether tosylate average Mn 900

Poly(ethylene glycol) methyl ether tosylate

Poly(ethylene glycol) methyl ether tosylate average Mn 900
Synonym(s):
Methoxy PEG tosylate, Methoxypolyethylene glycol tosylate, PEG tosylate, mPEG tosylate
Linear Formula:
CH3O(CH2CH2O)nCH2CH2OSO2C6H4CH3
CAS Number:

Products

Brand
Physical Form
Functional Group
Polymer Composition
Polymer Type