Skip to Content
Merck
Pitavastatin calcium ≥98% (HPLC)

Pitavastatin calcium

Pitavastatin calcium ≥98% (HPLC)
Synonym(s):
(+)monocalcium bis{(3R, 5S, 6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolyl]-3,5dihydroxy-6-heptenoate}, (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic acid calcium salt
Empirical Formula (Hill Notation):
C25H23FNO4 · 0.5Ca
CAS Number:
Molecular Weight:
440.49

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing