
Perfluorohexanoic acid

Synonym(s):
Undecafluorohexanoic acid, Perfluorocaproic acid, Perfluorohexanoic acid
Linear Formula:
CF3(CF2)4COOH
CAS Number:
Molecular Weight:
314.05
Products
Brand
Formula Weight
Manufacturer
Purity
Agency
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2