
Palmitic acid-1,2,3,4-13C4

Synonym(s):
Hexadecanoic acid-1,2,3,4-13C4
Linear Formula:
CH3(CH2)11(13CH2)313CO2H
CAS Number:
Molecular Weight:
260.39
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing