
Oxycodone Related Compound A

Synonym(s):
14-Hydroxycodeinone, (5α)-7,8-Didehydro-4,5-epoxy-14-hydroxy-3-methoxy-17-methylmorphinan-6-one
Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35
Products
Brand
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2