
Nateglinide impurity B

Synonym(s):
N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-L-phenylalanine
Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing