
Metazachlor Metabolite M479H016

Synonym(s):
3-[N-(2,6-Dimethylphenyl)-N-(1H-pyrazol-1-ylmethyl)aminocarbonylmethylsulfinyl]-2-hydroxypropanoic acid
Empirical Formula (Hill Notation):
C17H21N3O5S
CAS Number:
Molecular Weight:
379.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing