
Flavoxate Related Compound C

Synonym(s):
Ethyl 3-methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate, Ethyl 3-methylflavone-8-carboxylate
Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2