
Exemestane Related Compound C

Synonym(s):
1,4-Androstadiene-3,17-dione, 1-Dehydroandrostenedione, Androstadienedione
Empirical Formula (Hill Notation):
C19H24O2
CAS Number:
Molecular Weight:
284.39
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2