
Ethyl piperazinoacetate

Synonym(s):
1-(Ethoxycarbonylmethyl)piperazine, N-(Carboethoxymethyl)-piperazine, Ethyl 1-piperazineacetate
Empirical Formula (Hill Notation):
C8H16N2O2
Products
Brand
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing