![(E)-Metominostrobin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/779/846/deb2b0d4-8255-41e6-937f-7946e4715d4b/640/deb2b0d4-8255-41e6-937f-7946e4715d4b.png)
(E)-Metominostrobin
![(E)-Metominostrobin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/779/846/deb2b0d4-8255-41e6-937f-7946e4715d4b/640/deb2b0d4-8255-41e6-937f-7946e4715d4b.png)
Synonym(s):
Methyl (E)-α-methoxyimino-N-methyl-2-phenoxyphenylacetamide
Empirical Formula (Hill Notation):
C16H16N2O3
CAS Number:
Molecular Weight:
284.31
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1