![D-Ribulose 1,5-bisphosphate sodium salt hydrate ≥90% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/252/249/a490ca7c-6886-490d-a7c5-c4919e873483/640/a490ca7c-6886-490d-a7c5-c4919e873483.png)
D-Ribulose 1,5-bisphosphate sodium salt hydrate
![D-Ribulose 1,5-bisphosphate sodium salt hydrate ≥90% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/252/249/a490ca7c-6886-490d-a7c5-c4919e873483/640/a490ca7c-6886-490d-a7c5-c4919e873483.png)
Synonym(s):
D-erythro-2-Pentulose, 1,5-bis(dihydrogen phosphate), D-Ribulose 1,5-diphosphate, RuDP
Empirical Formula (Hill Notation):
C5H12O11P2 · xNa+ · yH2O
CAS Number:
Molecular Weight:
310.09 (anhydrous free acid basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing