DMT-5-Methyl-dC(ac) Phosphoramidite
Synonym(s):
5-Methyl-dC(ac) amidite
Empirical Formula (Hill Notation):
C42H52N5O8P
Molecular Weight:
785.86
Products
Product Category
Brand
Formula Weight
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing