(±)-CPP
Synonym(s):
(±)-3-(2-Carboxypiperazin-4-yl)propyl-1-phosphonic acid
Empirical Formula (Hill Notation):
C8H17N2O5P
CAS Number:
Molecular Weight:
252.20
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing