Skip to Content
Merck
(±)-Amphetamine-d5 (deuterium label on side chain) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

(±)-Amphetamine-d5 (deuterium label on side chain) solution

(±)-Amphetamine-d5 (deuterium label on side chain) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C9D5H8N
CAS Number:
Molecular Weight:
140.24

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique