
Diethyltoluamide

Synonym(s):
N,N-Diethyl-3-methylbenzamide, N,N-Diethyl-m-toluamide, m-Toluic acid diethylamide, DEET
Linear Formula:
CH3C6H4CON(C2H5)2
CAS Number:
Molecular Weight:
191.27
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2