![Cineole European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/341/464/c2116f21-43f3-45c3-a888-36026d5d95f0/640/c2116f21-43f3-45c3-a888-36026d5d95f0.png)
Cineole
![Cineole European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/341/464/c2116f21-43f3-45c3-a888-36026d5d95f0/640/c2116f21-43f3-45c3-a888-36026d5d95f0.png)
Synonym(s):
Eucalyptol, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing