
Cidofovir Related Compound A

Synonym(s):
1-[(S)-3-Hydroxy-2-(O-ethylphosphonomethoxy)propyl]cytosine
Empirical Formula (Hill Notation):
C10H18N3O6P
CAS Number:
Molecular Weight:
307.24
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing