Skip to Content
Merck
vibrant-m

Cefepime Related Compound E

vibrant-m
Synonym(s):
1-{[(6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-1-methylpyrrolidin-1-ium chloride
Empirical Formula (Hill Notation):
C13H20ClN3O3S
CAS Number:
Molecular Weight:
333.83

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade