
Boc-Ser(Bzl)-OH

Synonym(s):
N-(tert-Butoxycarbonyl)-O-benzyl-L-serine, Boc-O-benzyl-L-serine, Boc-Ser(Bzl)-OH
Linear Formula:
C6H5CH2OCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
295.33
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing