
Benzylpenicillin sodium

Synonym(s):
Penicillin G sodium salt, Benzylpenicillin sodium salt
Empirical Formula (Hill Notation):
C16H17N2NaO4S
CAS Number:
Molecular Weight:
356.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing