
Articaine hydrochloride

Synonym(s):
4-Methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylic acid methyl ester hydrochloride, Carticaine chlorhydrate, Carticaine hydrochloride, Articaine hydrochloride
Empirical Formula (Hill Notation):
C13H20N2O3S · HCl
CAS Number:
Molecular Weight:
320.84
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing