
α-Amylase from porcine pancreas

Synonym(s):
β-N-acetylglucosaminidase porcine placenta, PPA, al1,4 glucan-4-glucanohydrolase,, porcine pancreas α-amylase
Products
Brand
Sustainability
Biological Source
Feature
Physical Form
Tissue
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing