
4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate

Synonym(s):
6-Amino-2-mercapto-4-pyrimidinol monohydrate, 6-Amino-2-thiouracil monohydrate
Empirical Formula (Hill Notation):
C4H5N3OS · H2O
CAS Number:
Molecular Weight:
161.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing