
2H-Perfluoro-2-decenoic acid

Synonym(s):
3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Hexadecafluoro-2-decenoic acid
Empirical Formula (Hill Notation):
C10H2F16O2
CAS Number:
Molecular Weight:
458.10
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing