
2,2-Dimethyl-4-(1-methylethyl)-1,3-benzodioxole

Synonym(s):
2,2-Dimethyl-4-propan-2-yl-1,3-benzodioxole, 4-Isopropyl-2,2-dimethylbenzo[d][1,3]dioxole
Empirical Formula (Hill Notation):
C12H16O2
CAS Number:
Molecular Weight:
192.25
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1