
1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6- disulfonic acid trisodium salt

Synonym(s):
1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6- disulfonic acid trisodium salt
Empirical Formula (Hill Notation):
C16H9N2Na3O11S3
CAS Number:
Molecular Weight:
570.41
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing