Skip to Content
Merck
All Photos(1)

Documents

57492-U

Supelco

Supel Swift HLB SPE Tubes

weight 60 mg (bed), volume 3 mL, pk of 54 ea

Synonym(s):

polymeric general SPE cartidge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

description

detection: MS friendly

Quality Level

product line

Supel

packaging

pk of 54 ea

technique(s)

solid phase extraction (SPE): suitable

surface area

425-500 m2/g

volume

3 mL

weight

60 mg (bed)

particle size

40-75 μm (>90%)

pore size

80-210 Å pore size

Looking for similar products? Visit Product Comparison Guide

General description

Supel Swift HLB is a co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups. This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma). Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values. Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions. The product design allows for direct sample loading onto the sorbent bed, thereby eliminating conditioning of the cartridge. This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service