Z637386
Pursept®-AF disinfectant
volume 5 L
Synonym(s):
Disinfectant Solution
About This Item
Recommended Products
manufacturer/tradename
BRAND 44855
technique(s)
protein staining: suitable
volume
5 L
5000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Ingredients:
100 g Pursept-AF contain: 12.5 g Dicedyl-dimethyl-ammoniumchloride, 10.4 g Alkylpropylenediamine-bis-guanidiniumdiacetate, 1.5 g Bis(aminopropyl)-laurylamine, auxiliary agents
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service